Learn More
Invitrogen™ TIGIT Monoclonal Antibody (MBSA43), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
3305.00 € - 14910.00 €
Specifications
| Antigen | TIGIT |
|---|---|
| Clone | MBSA43 |
| Concentration | 1 mg/mL |
| Classification | Monoclonal |
| Conjugate | Functional Grade |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17141602
|
Invitrogen™
16-9500-38 |
5 mg |
3305.00 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17121602
|
Invitrogen™
16-9500-025 |
25 mg |
10000.00 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17131602
|
Affymetrix eBioscience
16-9500-050 |
50 mg | |||||||
Description
The MBSA43 MAb reacts with human T cell immunoreceptor with Ig and ITIM domains (TIGIT), which is also known as VSTM3 or WUCAM. This 26 kDa protein is a member of the poliovirus receptor (PVR) family. The expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but it is not expressed on B cells or naive CD4+ T cells. TIGIT may be upregulated on naive CD4+ T cells upon activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK and T cells with antigen presenting cells, fibroblasts, and endothelial cells that express PVR and PVR-like proteins. MBSA43 is reported to crossreact with Rhesus macaque.
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.Specifications
| TIGIT | |
| 1 mg/mL | |
| Functional Grade | |
| Mouse | |
| Human | |
| Q495A1 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| MBSA43 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 201633 | |
| Primary | |
| 4°C | |
| TIGIT |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.