missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIGIT Monoclonal Antibody (MBSA43), NovaFluor™ Red 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
182.00 € - 404.00 €
Specifications
| Antigen | TIGIT |
|---|---|
| Clone | MBSA43 |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30055573
|
Invitrogen™
H011T03R03-A |
100 Tests |
404.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30055324
|
Invitrogen™
H011T02R03-A |
25 Tests |
182.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIGIT Monoclonal antibody specifically detects TIGIT in Human samples. It is validated for Flow CytometrySpecifications
| TIGIT | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q495A1 | |
| 201633 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| MBSA43 | |
| Flow Cytometry | |
| NovaFluor Red 700 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
| IgG1 κ | |
| Affinity chromatography | |
| TIGIT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title