missing translation for 'onlineSavingsMsg'
Learn More

TIMM44 Antibody (0E2) [DyLight 680], Novus Biologicals Biologicals™

Product Code. 30522325 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522325 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522325 Supplier Novus Biologicals Supplier No. NBP343045FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TIMM44 Monoclonal antibody specifically detects TIMM44 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM44
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 0
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias MIMT44, mitochondrial import inner membrane translocase subunit TIM44, mitochondrial inner membrane translocase, TIM44DKFZp686H05241, translocase of inner mitochondrial membrane 44 homolog (yeast)
Host Species Mouse
Immunogen translocase of inner mitochondrial membrane 44 homolog(yeast) - (Uniprot# O43615)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10469
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.