missing translation for 'onlineSavingsMsg'
Learn More

Tiocarlide, LoGiCal

Product Code. 30463819
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30463819

Brand: LoGiCal LGCFOR3283.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Tiocarlide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tiocarlide
CAS 910-86-1
Analyte or Component Names Tiocarlide
Shipping Condition Room Temperature
Molecular Formula C23 H32 N2 O2 S
Certifications/Compliance ISO 9001
Synonym Thiourea, N,N'-bis[4-(3-methylbutoxy)phenyl]-, Carbanilide, 4,4'-bis(isopentyloxy)thio- (6 CI,7 CI,8 CI), 1,3-Bis[4-(isopentyloxy)phenyl]thiourea, 4,4'-Bis(isoamyloxy)thiocarbanilide, 4,4'-Bis(isopentyloxy)thiocarbanilide, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea, N,N'-Bis[p-(3-methylbutoxy)phenyl]thiourea
InChI Formula InChI=1 S/C23H32N2O2S/c1-17(2)13-15-26-21-9-5-19(6-10-21)24-23(28)25-20-7-11-22(12-8-20)27-16-14-18(3)4/h5-12,17-18 H,13-16H2,1-4H3,(H2,24,25,28)
SMILES CC(C)CCOc1ccc(NC(=S)Nc2ccc(OCCC(C)C)cc2)cc1
IUPAC Name 1,3-bis[4-(3-methylbutoxy)phenyl]thiourea
Industry Type Forensics
Molecular Weight (g/mol) 400.58
Quantity 10 mg
Formula Weight 400.2184 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.