missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiotropium Bromide Monohydrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Tiotropium bromide hydrate |
| CAS | 139404-48-1 |
| Analyte or Component Names | Tiotropium bromide hydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 N O4 S2 . Br . H2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Tiotropium Bromide Monohydrate, 3-Oxa-9-azatricyclo[3.3.1.02,4]nonane, 3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane deriv., Tiotropium bromide hydrate, (1alpha,2beta,4beta,5alpha,7beta)-7-[(Hydroxydi-2-thienylacetyl)oxy]-9,9-dimethyl-3-oxa-9-azoniatricyclo[3.3.1.02,4]nonane bromide hydrate |
| InChI Formula | InChI=1S/C19H22NO4S2.BrH.H2O/c1-20(2)12-9-11(10-13(20)17-16(12)24-17)23-18(21)19(22,14-5-3-7-25-14)15-6-4-8-26-15;;/h3-8,11-13,16-17,22H,9-10H2,1-2H3;1H;1H2/q+1;;/p-1/t11-,12-,13+,16-,17+;; |
| Solution Type | Neat |
| SMILES | O.[Br-].C[N+]1(C)[C@@H]2C[C@H](C[C@H]1[C@@H]3O[C@H]23)OC(=O)C(O)(c4cccs4)c5cccs5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?