missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIP120B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | TIP120B |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18617210
|
Novus Biologicals
NBP2-94561-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674301
|
Novus Biologicals
NBP2-94561-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIP120B Polyclonal antibody specifically detects TIP120B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TIP120B | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 23066 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cullin-associated and neddylation-dissociated 2 (putative), Cullin-associated and neddylation-dissociated protein 2, cullin-associated NEDD8-dissociated protein 2, epididymis tissue protein Li 169, KIAA0667TBP-interacting protein of 120 kDa B, p120 CAND2, TBP interacting protein, TBP-interacting protein 120B, TIP120BTp120b | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 850-1000 of human CAND2 (NP_001155971.1). VGQVAGPGHQRELKAVLLEALGSPSEDVRAAASYALGRVGAGSLPDFLPFLLEQIEAEPRRQYLLLHSLREALGAAQPDSLKPYAEDIWALLFQRCEGAEEGTRGVVAECIGKLVLVNPSFLLPRLRKQLAAGRPHTRSTVITAVKFLISD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title