Learn More
TLR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB100-56563
This item is not returnable.
View return policy
Description
TLR1 Polyclonal specifically detects TLR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot, Flow (Cell Surface), Flow (Intracellular).
Specifications
| Western Blot, Immunohistochemistry | |
| Protein G purified | |
| RUO | |
| Purified |
| CD281, CD281 antigen, DKFZp547I0610, DKFZp564I0682, KIAA0012MGC126311, MGC104956, MGC126312, rsc786, TIL, TIL. LPRS5, Toll/interleukin-1 receptor-like protein, toll-like receptor 1 | |
| 0.1 mg | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |