missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
241.00 € - 488.00 €
Specifications
| Classification | Polyclonal |
|---|---|
| Form | Purified |
| Target Species | Mouse, Rat |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18777603
|
Novus Biologicals
NB100-56563SS |
241.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18729433
|
Novus Biologicals
NB100-56563 |
0.1 mg |
488.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLR1 Polyclonal specifically detects TLR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot, Flow (Cell Surface), Flow (Intracellular).Specifications
| Polyclonal | |
| Purified | |
| Mouse, Rat | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title