Learn More
TLR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 13 publications
Brand: Novus Biologicals NB100-56563SS
This item is not returnable.
View return policy
Description
TLR1 Polyclonal specifically detects TLR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot, Flow (Cell Surface), Flow (Intracellular).
Specifications
| Polyclonal | |
| Mouse, Rat | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| Purified |