missing translation for 'onlineSavingsMsg'
Learn More

TLR4 Antibody (MTS510) - Azide and BSA Free, Novus Biologicals™

Product Code. 18740414 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.10mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18740414 0.1 mg 0.10mg
18118041 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18740414 Supplier Novus Biologicals Supplier No. NBP224865

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 11 publications

TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), CyTOF-ready, Block/Neutralize.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLR4
Applications Flow Cytometry, Blocking Assay, CyTOF
Classification Monoclonal
Clone MTS510
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot reported in scientific literature (PMID 23629653), Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22573811), Flow (Cell Surface) reported in scientific literature (PMID 15879150), CyTOF-ready, Block/Neutralize
Gene Accession No. O00206
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Gene Symbols TLR4
Host Species Rat
Immunogen This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors
Primary or Secondary Primary
Gene ID (Entrez) 7099
Test Specificity This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.