missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface), CyTOF-ready, Block/Neutralize.
Specifications
Specifications
| Antigen | TLR4 |
| Applications | Flow Cytometry, Blocking Assay, CyTOF |
| Classification | Monoclonal |
| Clone | MTS510 |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot reported in scientific literature (PMID 23629653), Flow Cytometry 1ul/1 million cells, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 22573811), Flow (Cell Surface) reported in scientific literature (PMID 15879150), CyTOF-ready, Block/Neutralize |
| Gene Accession No. | O00206 |
| Gene Alias | ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 |
| Gene Symbols | TLR4 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?