missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TLR8 Monoclonal antibody specifically detects TLR8 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
| Antigen | TLR8 |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Clone | 935166 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Flow Cytometry, CyTOF-ready |
| Formulation | 50mM Sodium Borate |
| Gene Alias | CD288, CD288 antigen, MGC119599, MGC119600, toll-like receptor 8 |
| Host Species | Mouse |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human TLR8, Glu27-Thr827, Accession # Q9NR97 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?