missing translation for 'onlineSavingsMsg'
Learn More

TLX3 Antibody [Janelia Fluor« 646], Novus Biologicals Biologicals™

Product Code. 30488740 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30488740 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30488740 Supplier Novus Biologicals Supplier No. NBP327888JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TLX3 Polyclonal antibody specifically detects TLX3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLX3
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias homeo box 11-like 2, HOX11L2MGC29804, RNXHomeobox protein Hox-11L2, T-cell leukemia homeobox 3, T-cell leukemia homeobox protein 3, T-cell leukemia, homeobox 3
Host Species Rabbit
Immunogen TLX3 antibody was raised against a 16 amino acid peptide near the amino terminus of human TLX3. The immunogen is located within the first 50 amino acids of TLX3.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30012
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.