missing translation for 'onlineSavingsMsg'
Learn More

TMEM106B Antibody (6H1) [mFluor Violet 500 SE], Novus Biologicals Biologicals™

Product Code. 30522364 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522364 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522364 Supplier Novus Biologicals Supplier No. NBP343046MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMEM106B Monoclonal antibody specifically detects TMEM106B in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM106B
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6H1
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias transmembrane protein 106B
Host Species Mouse
Immunogen transmembrane protein 106B - (Uniprot# Q9NUM4)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54664
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.