missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM41B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00527SS
This item is not returnable.
View return policy
Description
TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse samples. It is validated for Western Blot
Specifications
| TMEM41B | |
| Polyclonal | |
| Unconjugated | |
| Tris Glycine | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 1.0 mg/mL | |
| Western Blot 2 μg/mL | |
| KIAA0033MGC33897, MGC57262, transmembrane protein 41B | |
| Partial synthetic peptide made to an internal portion of the human TMEM41B protein (between amino acids 50-100) [UniProt Q5BJD5]. | |
| 0.025 mg | |
| Autophagy, Lipid and Metabolism | |
| 440026 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction