missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM41B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
218.00 € - 483.00 €
Specifications
| Antigen | TMEM41B |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18370882
|
Novus Biologicals
NBP3-00527 |
0.1 mg |
483.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622397
|
Novus Biologicals
NBP3-00527SS |
0.025 mg |
218.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMEM41B Polyclonal antibody specifically detects TMEM41B in Human, Mouse samples. It is validated for Western BlotSpecifications
| TMEM41B | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| KIAA0033MGC33897, MGC57262, transmembrane protein 41B | |
| Partial synthetic peptide made to an internal portion of the human TMEM41B protein (between amino acids 50-100) [UniProt Q5BJD5]. | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Autophagy, Lipid and Metabolism | |
| Tris Glycine | |
| 440026 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title