missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM97 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP1-30436SS
This item is not returnable.
View return policy
Description
TMEM97 Polyclonal specifically detects TMEM97 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TMEM97 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| anti, MAC30Protein MAC30, transmembrane protein 97 | |
| Rabbit | |
| 24 kDa | |
| 0.025 mL | |
| Cholesterol Metabolism, Ovarian Carcinoma Cell Markers | |
| 27346 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1-5 ug/ml, Immunohistochemistry-Paraffin 1:200 | |
| Q5BJF2, Q8VD00 | |
| TMEM97 | |
| Synthetic peptide made to an internal portion of the human TMEM97 protein (within residues 105-155. [Swiss-Prot: Q5BJF2] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted to react with monkey based on 100% sequence homology. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction