missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM97 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
244.00 € - 511.00 €
Specifications
| Antigen | TMEM97 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1-5 ug/ml, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18079223
|
Novus Biologicals
NBP1-30436SS |
0.025 mL |
244.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749973
|
Novus Biologicals
NBP1-30436 |
511.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TMEM97 Polyclonal specifically detects TMEM97 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TMEM97 | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1-5 ug/ml, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Rabbit | |
| Cholesterol Metabolism, Ovarian Carcinoma Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| anti, MAC30Protein MAC30, transmembrane protein 97 | |
| TMEM97 | |
| IgG | |
| Affinity Purified | |
| Human. Predicted to react with monkey based on 100% sequence homology. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q5BJF2, Q8VD00 | |
| 27346 | |
| Synthetic peptide made to an internal portion of the human TMEM97 protein (within residues 105-155. [Swiss-Prot: Q5BJF2] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 24 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title