Learn More
TNFRSF17 Rabbit, anti-Human, Clone: 5H7, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82855-1-RR-100UL
Description
The TNFRSF17 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TNFRSF17. This antibody recognizes human antigen. The TNFRSF17 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TNFRSF17 | |
| 5H7 | |
| B cell maturation protein, CD269, TNFRSF17 | |
| 100 μL | |
| 608 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |