missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF17 Rabbit, anti-Human, Clone: 5H7, Proteintech
Rabbit Recombinant Monoclonal Antibody
149.00 € - 439.00 €
Specifications
| Antigen | TNFRSF17 |
|---|---|
| Clone | 5H7 |
| Concentration | 300 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18026865
|
Proteintech
82855-1-RR-20UL |
20 μL |
149.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18094235
|
Proteintech
82855-1-RR-100UL |
100 μL |
439.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TNFRSF17 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TNFRSF17. This antibody recognizes human antigen. The TNFRSF17 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TNFRSF17 | |
| 300 μg/mL | |
| Liquid | |
| Human | |
| B cell maturation protein, CD269, TNFRSF17 | |
| Primary |
| 5H7 | |
| Western Blot, ELISA | |
| Rabbit | |
| 608 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title