missing translation for 'onlineSavingsMsg'
Learn More

Tolazoline Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30462019
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462019 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462019 Supplier LoGiCal Supplier No. LGCAMP0585.0001
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tolazoline Hydrochloride
CAS 59-97-2
Analyte or Component Names Tolazoline Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C10 H12 N2 . Cl H
Certifications/Compliance ISO 17034
Synonym Tolazoline Hydrochloride, 4,5-Dihydro-2-(phenylmethyl)-1H-imidazole hydrochloride, 2-Benzyl-2-imidazoline hydrochloride, 2-Benzyl-2-iminazoline hydrochloride, Benzazoline hydrochloride, 2-Benzyl-4,5-imidazoline hydrochloride, Phenylmethylimidazolin hydrochloride
InChI Formula InChI=1S/C10H12N2.ClH/c1-2-4-9(5-3-1)8-10-11-6-7-12-10;/h1-5H,6-8H2,(H,11,12);1H
Solution Type Single Solution
SMILES Cl.C(C1=NCCN1)c2ccccc2
IUPAC Name 2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 196.68
Quantity 1 mL
Formula Weight 196.0767 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.