missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TOP2A Recombinant Rabbit Monoclonal Antibody (24GB1915)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557792
This item is not returnable.
View return policy
Description
TOP2A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2 and M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.
Specifications
| TOP2A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TOP2A | |
| DNA gyrase; DNA topoisomerase (ATP-hydrolyzing); DNA topoisomerase 2-alpha; DNA topoisomerase 2-alpha-like protein; DNA Topoisomerase II alpha; DNA topoisomerase II, 170 kD; DNA topoisomerase II, alpha isozyme; EC 5.99.1.3; OTTHUMP00000207108; P11388-1; TOP2; Top-2; Top2a; TOPOII; topoisomerase (DNA) 2 alpha; topoisomerase (DNA) II alpha; topoisomerase (DNA) II alpha 170kDa; topoisomerase ii alpha; topoisomersae II; TP2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21973, 360243, 7153 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1915 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11388, P41516, Q01320 | |
| TOP2A | |
| A synthesized peptide derived from human Topoisomerase II alpha (aa 1500-1531). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction