missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TOP2A Recombinant Rabbit Monoclonal Antibody (24GB1915)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

198.00 € - 564.00 €

Specifications

Antigen TOP2A
Clone 24GB1915
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275023
GREENER_CHOICE
View Documents
Invitrogen™
MA557792
100 μL
564.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275989
View Documents
Invitrogen™
MA557793
20 μL
198.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TOP2A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2 and M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.
Specifications

Specifications

TOP2A
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
TOP2A
DNA gyrase; DNA topoisomerase (ATP-hydrolyzing); DNA topoisomerase 2-alpha; DNA topoisomerase 2-alpha-like protein; DNA Topoisomerase II alpha; DNA topoisomerase II, 170 kD; DNA topoisomerase II, alpha isozyme; EC 5.99.1.3; OTTHUMP00000207108; P11388-1; TOP2; Top-2; Top2a; TOPOII; topoisomerase (DNA) 2 alpha; topoisomerase (DNA) II alpha; topoisomerase (DNA) II alpha 170kDa; topoisomerase ii alpha; topoisomersae II; TP2A
TOP2A
Primary
Affinity chromatography
24GB1915
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P11388, P41516, Q01320
21973, 360243, 7153
A synthesized peptide derived from human Topoisomerase II alpha (aa 1500-1531).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.