missing translation for 'onlineSavingsMsg'
Learn More

Topiramate, Lipomed

Product Code. 30462279
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462279 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462279 Supplier Lipomed Supplier No. LPMTPR139150
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Topiramate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Topiramate
CAS 97240-79-4
Analyte or Component Names Topiramate
Shipping Condition Room Temperature
Molecular Formula C12 H21 N O8 S
Synonym Topiramate, Epitoma, Epitomax, McN 4853, RWJ 17021, 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv., 5 H-Bis[1,3 ]dioxolo[4,5-b:4',5'-d]pyran, beta-D-fructopyranose deriv.
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC1(C)O[C@@H]2 CO[C@@]3(COS(=O)(=O)N)OC(C)(C)O[C@H]3 [C@@H]2O1
IUPAC Name [(3 AS,5 aR,8 aR,8 bS)-2,2,7,7-tetramethyl-5,5 A,8 A,8 b-tetrahydrodi[1,3 ]dioxolo[5,4-b:5',3'-d]pyran-3 A-yl]methyl sulfamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 339.36
Quantity 50 mg
Formula Weight 339.0988 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.