missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Topoisomerase I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30961-25ug
This item is not returnable.
View return policy
Description
Topoisomerase I Polyclonal antibody specifically detects Topoisomerase I in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Topoisomerase I | |
| Polyclonal | |
| Immunohistochemistry-Paraffin : 5-10 μg/mL | |
| DNA topoisomerase 1, DNA topoisomerase I, EC 5.99.1.2, TOPI, topoisomerase (DNA) I, type I DNA topoisomerase | |
| Synthetic peptide made to human Topoisomerase I | |
| 25 μg | |
| Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 7150 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction