missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Topoisomerase I Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | Topoisomerase I |
|---|---|
| Dilution | Immunohistochemistry-Paraffin : 5-10 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067203
|
Novus Biologicals™
NBP3-30961-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067074
|
Novus Biologicals™
NBP3-30961-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Topoisomerase I Polyclonal antibody specifically detects Topoisomerase I in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Topoisomerase I | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing | |
| Phosphate buffered saline | |
| 7150 | |
| IgG | |
| Antigen Affinity-purified |
| Immunohistochemistry-Paraffin : 5-10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA topoisomerase 1, DNA topoisomerase I, EC 5.99.1.2, TOPI, topoisomerase (DNA) I, type I DNA topoisomerase | |
| Synthetic peptide made to human Topoisomerase I | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title