missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Topoisomerase I Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-30961-100ug

 View more versions of this product

Product Code. 30067203

  • 508.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Topoisomerase I Polyclonal antibody specifically detects Topoisomerase I in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Topoisomerase I
Polyclonal
Immunohistochemistry-Paraffin : 5-10 μg/mL
DNA topoisomerase 1, DNA topoisomerase I, EC 5.99.1.2, TOPI, topoisomerase (DNA) I, type I DNA topoisomerase
Synthetic peptide made to human Topoisomerase I
100 μg
Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing
7150
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunohistochemistry (Paraffin)
Unconjugated
Phosphate buffered saline
Rabbit
Antigen Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.