Learn More
Invitrogen™ TPP2 Recombinant Rabbit Monoclonal Antibody (24GB2015)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557799
 View more versions of this product
Product Code. 30275630
  • 198.00 € / 20µL
Description
TPP2 Recombinant Monoclonal Antibody for Western Blot
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TPP2
Recombinant Monoclonal
Unconjugated
TPP2
TPP2; TPP-2; TppII; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase 2; tripeptidyl peptidase II; tripeptidyl-peptidase 2; Tripeptidyl-peptidase II
Rabbit
Affinity chromatography
RUO
22019, 7174, 81815
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2015
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P29144, Q64514, Q64560
TPP2
A synthesized peptide derived from human Tripeptidyl peptidase II (aa 900-1150).
20 μL
Primary
Human, Mouse, Rat