missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Trabikibart Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558863
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Trabikibart, also known as CSL311, is a fully human monoclonal antibody that targets the common beta chain (beta c) receptor, shared by the interleukin-3 (IL-3), granulocyte-macrophage colony-stimulating factor (GM-CSF), and interleukin-5 (IL-5) cytokines. Colony stimulating factor 2 receptor subunit beta (CSF2RB), also known as CD131, is a critical component of the cytokine receptor family involved in hematopoiesis and immune response regulation. Located on chromosome 22q13.1, the CSF2RB gene encodes the common beta-subunit that pairs with the alpha-subunits of receptors for granulocyte-macrophage colony-stimulating factor (GM-CSF), interleukin-3 (IL-3), and interleukin-5 (IL-5). Structurally, the beta-subunit contributes to the formation of high-affinity receptor complexes essential for signal transduction. Functionally, upon ligand binding, CSF2RB stimulates various intracellular signaling cascades that mediate cellular proliferation, differentiation, and survival, particularly of hematopoietic progenitor cells and immune cells. Aberrations in CSF2RB expression or function can lead to immune dysfunctions and are associated with diseases such as asthma and leukemia. Recent studies have highlighted the importance of the CSF2RB in regulatory T cells, underscoring its role in autoimmune disease prevention.
Specifications
| Trabikibart | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction