missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody (12A380) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-56748
This item is not returnable.
View return policy
Description
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface).
Specifications
| TRANCE/TNFSF11/RANK L | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 4-6 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 14726437), Immunohistochemistry-Paraffin 1:50 - 1:250, Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (Yoneda et al (2004)), Flow (Cell Surface) reported in scientific literature (PMID 14726437) | |
| TNFSF11 | |
| A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen. | |
| 0.1 mg | |
| Cancer | |
| 8600 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 12A380 | |
| Unconjugated | |
| CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction