missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRANCE/TNFSF11/RANK L Antibody (12A380) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
226.00 € - 521.00 €
Specifications
| Antigen | TRANCE/TNFSF11/RANK L |
|---|---|
| Clone | 12A380 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 4-6 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 14726437), Immunohistochemistry-Paraffin 1:50 - 1:250, Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (Yoneda et al (2004)), Flow (Cell Surface) reported in scientific literature (PMID 14726437) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|
18709453
|
Novus Biologicals
NB100-56748SS |
0.025 mg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18757293
|
Novus Biologicals
NB100-56748 |
0.1 mg |
521.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
TRANCE/TNFSF11/RANK L Monoclonal specifically detects TRANCE/TNFSF11/RANK L in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface).Specifica
| TRANCE/TNFSF11/RANK L | |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| CD254, CD254 antigen, OPGLOPTB2, Osteoclast differentiation factor, Osteoprotegerin ligand, RANKLreceptor activator of nuclear factor kappa B ligand, Receptor activator of nuclear factor kappa-B ligand, sOdf, TNF-related activation-induced cytokine, TRANCEODFhRANKL2, tumor necrosis factor (ligand) superfamily, member 11, tumor necrosis factor ligand superfamily member 11 | |
| TNFSF11 | |
| IgG1 κ | |
| Protein G purified |
| 12A380 | |
| Western Blot 4-6 ug/ml, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 14726437), Immunohistochemistry-Paraffin 1:50 - 1:250, Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (Yoneda et al (2004)), Flow (Cell Surface) reported in scientific literature (PMID 14726437) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 8600 | |
| A bacterially expressed fusion protein containing amino acid residues 1-317 of mouse TRANCE was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto