missing translation for 'onlineSavingsMsg'
Learn More

trans-Cinnamaldehyde, 99%

Product Code. 10459400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
500g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10459400 500 g Glass bottle 500g
10020820 50 g Glass bottle 50g
10560011 1 kg Glass bottle 1kg
11914781 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10459400 Supplier Thermo Scientific Acros Supplier No. 110355000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 14371-10-9 | C9H8O | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14371-10-9
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.16
MDL Number MFCD00007000
InChI Key KJPRLNWUNMBNBZ-QPJJXVBHSA-N
Synonym cinnamaldehyde, trans-cinnamaldehyde, cinnamic aldehyde, e-cinnamaldehyde, cinnamal, 3-phenylacrylaldehyde, zimtaldehyde, cinnamylaldehyde, 3-phenylpropenal, 2e-3-phenylprop-2-enal
PubChem CID 637511
ChEBI CHEBI:16731
SMILES O=C\C=C\C1=CC=CC=C1

Specifications

Melting Point -7.5°C
Color Yellow
Density 1.0500g/mL
Boiling Point 250.0°C to 252.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCHO
Refractive Index 1.6210 to 1.6230
Quantity 500 g
Beilstein 07,348
Merck Index 14,2297
Specific Gravity 1.05
Solubility Information Solubility in water: 1.1g/L (20°C) (kolbenmethode).
IUPAC Name (E)-3-phenylprop-2-enal
Formula Weight 132.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material trans-Cinnamaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • trans-Cinnamaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: trans-Cinnamaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.