missing translation for 'onlineSavingsMsg'
Learn More

transgelin 2 Antibody (7D0) [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30522469 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30522469 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30522469 Supplier Novus Biologicals Supplier No. NBP343050APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

transgelin 2 Monoclonal antibody specifically detects transgelin 2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen transgelin 2
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7D0
Conjugate APC
Formulation PBS
Gene Alias KIAA0120HA1756, SM22-alpha homolog, transgelin 2, transgelin-2
Host Species Mouse
Immunogen transgelin 2 - (Uniprot# P37802)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8407
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.