missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human EGFR / ErbB1 / HER1 siRNA
Blocks transcription and translation of EGFR / ErbB1 / HER1 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_005219 |
---|---|
Cell Lines Tested | WT-EGFR-CHO EGFR overexpressing CHO cells and MDA-MB 468 Human Breast Cancer Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | EGFR |
Gene ID (Entrez) | 1956 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019181
|
enQuireBio™
QX18-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009181
|
enQuireBio™
QX18-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005219 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
1956 | |
ERBB1, HER1, ERBB, mENA, PIG61, NISBD2, Epidermal growth factor receptor, EGF-R | |
Lyophilized |
WT-EGFR-CHO EGFR overexpressing CHO cells and MDA-MB 468 Human Breast Cancer Cells. | |
EGFR | |
siRNA | |
Human |