missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat Nerve growth factor receptor / p75NTR siRNA
Blocks transcription and translation of Nerve growth factor receptor / p75NTR from Rat.
250.00 € - 369.00 €
Specifications
Accession Number | NP_036742 |
---|---|
Cell Lines Tested | Pc12 Rat Pheocromocytoma Cells from the Adrenal Medulla and Rat Primary Neurons. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Ngfr |
Gene ID (Entrez) | 24596 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099211
|
enQuireBio™
QX35-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089211
|
enQuireBio™
QX35-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036742 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
24596 | |
p75NTR, Tnfrsf16, p75, RNNGFRR, LNGFR, low affinity nerve growth factor receptor, Tumor necrosis factor receptor superfamily member 16, NGFR | |
Lyophilized |
Pc12 Rat Pheocromocytoma Cells from the Adrenal Medulla and Rat Primary Neurons. | |
Ngfr | |
siRNA | |
Rat |