missing translation for 'onlineSavingsMsg'
Learn More

TRAPPC9 Antibody (8A2) [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30519484 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519484 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519484 Supplier Novus Biologicals Supplier No. NBP343052R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TRAPPC9 Monoclonal antibody specifically detects TRAPPC9 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRAPPC9
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8A2
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias IKBKBBP, IKK2 binding protein, KIAA1882IBP, MRT13MGC4769, NIBPMGC4737, NIK- and IKBKB-binding protein, NIK and IKK-beta binding protein, T1, trafficking protein particle complex 9, trafficking protein particle complex subunit 9, TRAPP 120 kDa subunit, TRS120, Tularik gene 1 protein
Host Species Mouse
Immunogen trafficking protein particle complex 9 - (Uniprot# Q96Q05)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 83696
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.