Learn More
TREK1/KCNK2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26807-1-AP-20UL
Description
The TREK1/KCNK2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TREK1/KCNK2. This antibody recognizes human antigen. The TREK1/KCNK2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TREK1/KCNK2 | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| hTREK 1c, hTREK 1e, K2p2.1, KCNK2, TPKC1, TREK, TREK 1, TREK 1 K(+) channel subunit, TREK1 | |
| 20 μL | |
| 3776 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.