missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREK1/KCNK2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | TREK1/KCNK2 |
|---|---|
| Concentration | 450 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18060616
|
Proteintech
26807-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18048205
|
Proteintech
26807-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TREK1/KCNK2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TREK1/KCNK2. This antibody recognizes human antigen. The TREK1/KCNK2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TREK1/KCNK2 | |
| Western Blot, ELISA | |
| Rabbit | |
| 3776 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 450 μg/mL | |
| Liquid | |
| Human | |
| hTREK 1c, hTREK 1e, K2p2.1, KCNK2, TPKC1, TREK, TREK 1, TREK 1 K(+) channel subunit, TREK1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title