missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone acetate, 1mg/ml in Isopropanol, Lipomed

Product Code. 30457935
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457935 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457935 Supplier Lipomed Supplier No. LPMTBL15281LI
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Trenbolone acetate, 1mg/ml in Isopropanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trenbolone Acetate
CAS 10161-34-9
Analyte or Component Names Trenbolone Acetate
Shipping Condition Room Temperature
Molecular Formula C20 H24 O3
Concentration 1 mg/mL
Synonym Estra-4,9,11-trien-3-one, 17β-hydroxy-, acetate (7 CI,8 CI), (17β)-17-(Acetyloxy)estra-4,9,11-trien-3-one, 17β-Acetoxy-Δ4,9,11-estratrien-3-one, 17β-Acetoxyestra-4,9,11-trien-3-one, 17β-Hydroxyestra-4,9,11-trien-3-one 17-acetate, 3-Oxo-17β-acetoxyestra-4,9,11-triene, Component T-H, Component T-S, Finaplix, Finaplix H, Finaplix-S, RU 1697, Trenbolone acetate, Trienbolone acetate
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CCC4=CC(=O)CCC4=C3C=C[C@]12 C
IUPAC Name [(8 S,13 S,14 S,17 S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1 H-cyclopenta[a]phenanthren-17-yl] acetate
Solvent or Matrix Isopropanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 312.4
Quantity 1 mL
Formula Weight 312.1725 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.