missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone acetate, Lipomed

Product Code. 30460494
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460494 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460494 Supplier Lipomed Supplier No. LPMTBL1528100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Trenbolone acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trenbolone Acetate
CAS 10161-34-9
Analyte or Component Names Trenbolone Acetate
Shipping Condition Ice Pk. (-20°C)
Molecular Formula C20 H24 O3
Synonym Estra-4,9,11-trien-3-one, 17β-hydroxy-, acetate (7 CI,8 CI), (17β)-17-(Acetyloxy)estra-4,9,11-trien-3-one, 17β-Acetoxy-Δ4,9,11-estratrien-3-one, 17β-Acetoxyestra-4,9,11-trien-3-one, 17β-Hydroxyestra-4,9,11-trien-3-one 17-acetate, 3-Oxo-17β-acetoxyestra-4,9,11-triene, Component T-H, Component T-S, Finaplix, Finaplix H, Finaplix-S, RU 1697, Trenbolone acetate, Trienbolone acetate
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CCC4=CC(=O)CCC4=C3C=C[C@]12 C
IUPAC Name [(8 S,13 S,14 S,17 S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1 H-cyclopenta[a]phenanthren-17-yl] acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 312.4
Quantity 100 mg
Formula Weight 312.1725 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.