missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate), LoGiCal

Product Code. 30457793
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457793 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457793 Supplier LoGiCal Supplier No. LGCFOR3717.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trenbolone Cyclohexylmethylcarbonate
CAS 23454-33-3
Analyte or Component Names Trenbolone Cyclohexylmethylcarbonate
Shipping Condition Room Temperature
Molecular Formula C26 H34 O4
Certifications/Compliance ISO 9001
Synonym Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate), Estra-4,9,11-trien-3-one, 17-[[(cyclohexylmethoxy)carbonyl]oxy]-, (17β)-, Estra-4,9,11-trien-3-one, 17β-hydroxy-, cyclohexylmethyl carbonate (8CI), Carbonic acid, cyclohexylmethyl ester, ester with 17β-hydroxyestra-4,9,11-trien-3-one (8CI), Cyclohexanemethanol, hydrogen carbonate, ester with 17β-hydroxyestra-4,9,11-trien-3-one (8CI), Parabolan, RU 2580, Trenbolone cyclohexylmethyl carbonate, Trenbolone hexahydrobenzyl carbonate
InChI Formula InChI=1S/C26H34O4/c1-26-14-13-21-20-10-8-19(27)15-18(20)7-9-22(21)23(26)11-12-24(26)30-25(28)29-16-17-5-3-2-4-6-17/h13-15,17,22-24H,2-12,16H2,1H3/t22-,23+,24+,26+/m1/s1
Solution Type Neat
SMILES C[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@H]2OC(=O)OCC5CCCCC5
IUPAC Name cyclohexylmethyl [(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl] carbonate
Industry Type Food Testing
Molecular Weight (g/mol) 410.55
Quantity 10 mg
Formula Weight 410.554 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.