missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trenbolone Enantate, Mikromol™
Description
Trenbolone Enantate
Specifications
Specifications
| Chemical Name or Material | Trenbolone enanthate |
| CAS | 1629618-98-9 |
| Analyte or Component Names | Trenbolone enanthate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H34 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (17β)-17-[(1-Oxoheptyl)oxy]-estra-4,9,11-trien-3-one, Trenbolone Enanthate |
| InChI Formula | InChI=1S/C25H34O3/c1-3-4-5-6-7-24(27)28-23-13-12-22-21-10-8-17-16-18(26)9-11-19(17)20(21)14-15-25(22,23)2/h14-16,21-23H,3-13H2,1-2H3/t21-,22+,23+,25+/m1/s1 |
| SMILES | CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3C=C[C@]12C |
| IUPAC Name | [(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl] heptanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?