missing translation for 'onlineSavingsMsg'
Learn More

Tricaprin 8000 μg/mL in Pyridine, Dr. Ehrenstorfer

Product Code. 30321934
Click to view available options
Quantity:
5 x 5 mL
Unit Size:
25mL
This item is not returnable. View return policy

Product Code. 30321934

Brand: Dr. Ehrenstorfer DRESY09010004PY

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tricaprin
CAS 621-71-6
Analyte or Component Names Tricaprin
Shipping Condition Room Temperature
Molecular Formula C33 H62 O6
Certifications/Compliance ISO 17034
Synonym Decanoic acid 1,1',1''-(1,2,3-propanetriyl) ester, tri-Caprin, Decanoic acid 1,2,3-propanetriyl ester, tri-Decanoin, Actor M 2, Capric acid triglyceride, Capric triglyceride, Captex 1000, Decanoic acid triglyceride, Dynasan 110, Glycerin tricaprate, Glycerin tridecanoate, Glycerol tricaprate, Glycerol tricaprin, Glycerol tridecanoate, Glyceryl tricaprate, NSC 147475, Panacet 1000, Tri(decanoyl)glycerol, Tricapric glyceride, Tricaprin, Tridecanoin, Triglyceride DDD
InChI Formula InChI=1S/C33H62O6/c1-4-7-10-13-16-19-22-25-31(34)37-28-30(39-33(36)27-24-21-18-15-12-9-6-3)29-38-32(35)26-23-20-17-14-11-8-5-2/h30H,4-29H2,1-3H3
SMILES CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC
IUPAC Name 2,3-di(decanoyloxy)propyl decanoate
Solvent or Matrix Pyridine
Industry Type Food Testing
Molecular Weight (g/mol) 554.84
Quantity 5 x 5 mL
Formula Weight 554.4546 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.