missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRIF/TICAM1 Polyclonal specifically detects TRIF/TICAM1 in Mouse, Rat, Porcine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated.
Specifications
Specifications
| Antigen | TRIF/TICAM1 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot |
| Concentration | 1.0 mg/ml |
| Host Species | Rabbit |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Target Species | Pig |
| Content And Storage | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Form | Antisera |
| Isotype | IgG |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?