missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIF/TICAM1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 28 publications
230.00 € - 552.00 €
Specifications
| Antigen | TRIF/TICAM1 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1 - 2 ug/ml, Simple Western 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunoprecipitation reported in scientific literature (PMID 28007523), Immunohistochemistry-Paraffin 1:200, Immunoblotting reported in scientific literature (PMID 27043414), KnockDown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18765043
|
Novus Biologicals
NB120-13810SS |
0.025 mg |
230.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18768413
|
Novus Biologicals
NB120-13810 |
0.1 mg |
552.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TRIF/TICAM1 Polyclonal specifically detects TRIF/TICAM1 in Mouse, Rat, Porcine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated.Specifications
| TRIF/TICAM1 | |
| Western Blot 1 - 2 ug/ml, Simple Western 1:100, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunoprecipitation reported in scientific literature (PMID 28007523), Immunohistochemistry-Paraffin 1:200, Immunoblotting reported in scientific literature (PMID 27043414), KnockDown Validated | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse, Rat, Pig | |
| Q80UF7 | |
| 148022 | |
| Synthetic peptide made to an internal portion of the mouse TRIF protein (between amino acids 100-180) [UniProt Q80UF7]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 76 kDa |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoblot, KnockDown | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta | |
| TICAM1 | |
| IgG | |
| Immunogen affinity purified | |
| The will cross-react with human TRIF, but additional bands may be seen. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title