missing translation for 'onlineSavingsMsg'
Learn More

Trifluoperazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30459406
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459406 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459406 Supplier LoGiCal Supplier No. LGCAMP0441.0001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Trifluoperazine Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifluoperazine hydrochloride
CAS 440-17-5
Analyte or Component Names Trifluoperazine hydrochloride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C21 H24 F3 N3 S . 2 Cl H
Certifications/Compliance ISO 17034
Synonym Trifluoperazine hydrochloride, 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)-10 H-phenothiazine dihydrochloride, 10 H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, hydrochloride (1:2)10 H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (9 CI), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (6 CI,8 CI), 2-Trifluoromethyl-10-[3-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride
InChI Formula InChI=1 S/C21H24F3N3S0.2 ClH/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)28-20-8-7-16(15-18(20)27)21(22,23)24;;/h2-3,5-8,15 H,4,9-14H2,1H3;2*1 H
SMILES Cl.Cl.CN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1
IUPAC Name 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 480.42
Quantity 1 mL
Formula Weight 479.1177 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.