missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoperazine Hydrochloride, Mikromol™
Description
Trifluoperazine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Trifluoperazine hydrochloride |
| CAS | 440-17-5 |
| Analyte or Component Names | Trifluoperazine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H24 F3 N3 S . 2 Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Trifluoperazine hydrochloride, 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)-10H-phenothiazine dihydrochloride, 10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, hydrochloride (1:2)10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (9CI), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (6CI,8CI), 2-Trifluoromethyl-10-[3-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, Eskazine, Eskazinyl, Fluoperazine, Jatroneural, Modalina, SKF 5019, Stelazine, TFP, Terfluzine, Trazine, Trifluoperazine dihydrochloride, Trifluoperazine hydrochloride, Trifluoroperazine dihydrochloride, Trifluoroperazine hydrochloride, Triftazin, Triphthasine, Triphthazine, Triphthazine dihydrochloride, Tryptazine, Tryptazine dihydrochloride, Stelazine, SKF-5019-A2 |
| InChI Formula | InChI=1S/C21H24F3N3S.2ClH/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)28-20-8-7-16(15-18(20)27)21(22,23)24;;/h2-3,5-8,15H,4,9-14H2,1H3;2*1H |
| SMILES | Cl.Cl.CN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?