missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trilostane, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Trilostane |
| CAS | 13647-35-3 |
| Analyte or Component Names | Trilostane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H27 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Desopan, Modrastane, Modrefen, Modrenal, Trilostane, Vetoryl, Win 24540, Androst-2-ene-2-carbonitrile, 4,5-epoxy-3,17-dihydroxy-, (4alpha,5alpha,17beta)- |
| InChI Formula | InChI=1S/C20H27NO3/c1-18-7-6-14-12(13(18)3-4-15(18)22)5-8-20-17(24-20)16(23)11(10-21)9-19(14,20)2/h12-15,17,22-23H,3-9H2,1-2H3/t12-,13-,14-,15-,17+,18-,19+,20+/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@]12CC[C@H]3[C@@H](CC[C@@]45O[C@@H]4C(=C(C[C@]35C)C#N)O)[C@@H]1CC[C@@H]2O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction