missing translation for 'onlineSavingsMsg'
Learn More

TRIM27 Antibody (PCRP-TRIM27-1B3), Biotin, R&D Systems™

Product Code. 18607746 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18607746 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18607746 Supplier Novus Biologicals Supplier No. NBP308579B
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

TRIM27 Monoclonal specifically detects TRIM27 in Human samples. It is validated for Flow Cytometry/Immunocytochemistry/Immunofluorescence/Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIM27
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-TRIM27-1B3
Conjugate Biotin
Formulation PBS
Gene Alias Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP
Host Species Mouse
Immunogen Recombinant full-length human TRIM27 protein (Uniprot: P14373)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5987
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.