missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM29 Antibody (TRIM29/9256R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23806-20ug
This item is not returnable.
View return policy
Description
TRIM29 Monoclonal antibody specifically detects TRIM29 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TRIM29 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Ataxia telangiectasia group D-associated protein, ATDC, ATDC ataxia-telangiectasia group D-associated protein, FLJ36085, tripartite motif containing 29, tripartite motif protein TRIM29, tripartite motif-containing 29, tripartite motif-containing protein 29 | |
| Recombinant fragment (around aa 1-200) of human TRIM29 protein (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| TRIM29/9256R | |
| Unconjugated | |
| 1 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 23650 | |
| Store at 4°C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction